Nonanal dimethyl acetal ≥97.0% (by GC)

Supplier: TCI America

N0915-25G N0915-5G
TCN0915-25GEA 107.44 CAD
TCN0915-25G TCN0915-5G
Nonanal dimethyl acetal ≥97.0% (by GC)
Nonanal dimethyl acetal

CAS Number: 18824-63-0
MDL Number: MFCD00036646
Molecular Formula: C11H24O2
Molecular Weight: 188.31
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Specific Gravity (20/20): 0.85


Other Product Names / Synonyms: 1,1-Dimethoxynonane, Nonyl Aldehyde Dimethyl Acetal, Pelargonaldehyde Dimethyl Acetal

Formula: C₁₁H₂₄O₂
Boiling Pt: 88 °C
Density: 0.85 (20 °C)
MDL Number: MFCD00036646
CAS Number: 18824-63-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d0fa72629c5550ed6c8371eae3a7bf764e4b924&vid=1d0fa72629c5550ed6c8371eae3a7bf764e4b924&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR