5-Norbornene-2-carboxamide (mixture of isomers) ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

N0911-25G N0911-5G
TCN0911-25GEA 148.52 CAD
TCN0911-25G TCN0911-5G
5-Norbornene-2-carboxamide (mixture of isomers) ≥98.0% (by HPLC, total nitrogen)
5-Norbornene-2-carboxamide (mixture of isomers)

CAS Number: 95-17-0
MDL Number: MFCD00187896
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 188


Other Product Names / Synonyms: Bicyclo[2.2.1]hept-5-ene-2-carboxamide

Formula: C₈H₁₁NO
Melting Pt: 188 °C
MDL Number: MFCD00187896
CAS Number: 95-17-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=185834dc4de7519bc96719ddebccedd9006c9a53&vid=185834dc4de7519bc96719ddebccedd9006c9a53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR