3-Nitro-L-tyrosine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
N0905-25G
N0905-5G
TCN0905-25GEA
1031.74
CAD
TCN0905-25G
TCN0905-5G
3-Nitro-L-tyrosine ≥98.0% (by HPLC, titration analysis)
3-Nitro-L-tyrosine
CAS Number: 621-44-3
MDL Number: MFCD00007123
Molecular Formula: C9H10N2O5
Molecular Weight: 226.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 224
Specific rotation [a]20/D: 3 deg (C=1, 1mol/L HCl)
Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Tyr(3-NO2)-OH
Formula:
C₉H₁₀N₂O₅ Melting Pt: 224 °C |
MDL Number:
MFCD00007123 CAS Number: 621-44-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...