Nisoldipine ≥98.0%

Supplier: TCI America

Synonyms: Isobutyl Methyl 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylate, 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic Acid Isobutyl Methyl Ester, 3-Isobutyl 5-methyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate

N0900-1G N0900-5G
TCN0900-1GEA 150.42 CAD
TCN0900-1G TCN0900-5G
Nisoldipine ≥98.0%
Nisoldipine

CAS Number: 63675-72-9
MDL Number: MFCD00478055
Molecular Formula: C20H24N2O6
Molecular Weight: 388.42
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 153


Product Categories: Bioscience, Antihypertensive Ingredients, Voltage-dependent Calcium Channel (VDCC) Antagonists
Other Product Names / Synonyms: 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic Acid Isobutyl Methyl Ester, Isobutyl Methyl 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylate

Formula: C₂₀H₂₄N₂O₆
MW: 388.42 g/mol
Melting Pt: 153 °C
MDL Number: MFCD00478055
CAS Number: 63675-72-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0d3574e4bb307e9dcd9e33b93fa7016b08c85cd&vid=d0d3574e4bb307e9dcd9e33b93fa7016b08c85cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR