Nilvadipine ≥95.0% (by total nitrogen basis)

Supplier: TCI America

N0899-1G N0899-5G
TCN0899-1GEA 172.22 CAD
TCN0899-1G TCN0899-5G
Nilvadipine ≥95.0% (by total nitrogen basis)
Nilvadipine

CAS Number: 75530-68-6
MDL Number: MFCD00867065
Molecular Formula: C19H19N3O6
Molecular Weight: 385.38
Purity/Analysis Method: >95.0% (N)
Form: Crystal
Melting point (°C): 151


Product Categories: Bioscience, Antihypertensive Ingredients
Other Product Names / Synonyms: 2-Cyano-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 5-Isopropyl 3-Methyl Ester, 5-Isopropyl 3-Methyl 2-Cyano-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate

Formula: C₁₉H₁₉N₃O₆
Melting Pt: 151 °C
MDL Number: MFCD00867065
CAS Number: 75530-68-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15b6530c74c787d9689bda3b3a3788393262aa4a&vid=15b6530c74c787d9689bda3b3a3788393262aa4a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR