L-Norvaline ethyl ester hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: (S)-2-Aminovaleric Acid Ethyl Ester Hydrochloride, H-Nva-OEt · HCl, Ethyl (S)-2-Aminovalerate Hydrochloride, Ethyl (S)-2-Aminopentanoate Hydrochloride, H-Nva-OEt.HCl, (S)-2-Aminopentanoic Acid Ethyl Ester Hydrochloride

N0898-25G N0898-5G
TCN0898-25GEA 176.96 CAD
TCN0898-25G TCN0898-5G
L-Norvaline ethyl ester hydrochloride ≥98.0%
L-Norvaline ethyl ester hydrochloride

CAS Number: 40918-51-2
MDL Number: MFCD08458628
Molecular Formula: C7H15NO2
Molecular Weight: 181.66
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 109
Specific rotation [a]20/D: 9 deg (C=2, H2O)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: (S)-2-Aminopentanoic Acid Ethyl Ester Hydrochloride, (S)-2-Aminovaleric Acid Ethyl Ester Hydrochloride, Ethyl (S)-2-Aminopentanoate Hydrochloride, Ethyl (S)-2-Aminovalerate Hydrochloride, H-Nva-OEt.HCl

Formula: C₇H₁₅NO₂
MW: 181.66 g/mol
Melting Pt: 109 °C
MDL Number: MFCD08458628
CAS Number: 40918-51-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce25a264e1d35ff41a1bc59effd432c57113e979&vid=ce25a264e1d35ff41a1bc59effd432c57113e979&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR