5-Norbornene-2-carbonitrile (mixture of isomers) ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Bicyclo[2.2.1]hept-5-ene-2-carbonitrile, 5-Norbornene-2-carbonitrile, 5-Cyanobicyclo[2.2.1]hept-2-ene, 5-Cyano-2-norbornene
CAS Number: 95-11-4
MDL Number: MFCD00167563
Molecular Formula: C8H9N
Molecular Weight: 119.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Melting point (°C): 10
Flash Point (°C): 66
Specific Gravity (20/20): 1.01
Other Product Names / Synonyms: Bicyclo[2.2.1]hept-5-ene-2-carbonitrile, 5-Cyanobicyclo[2.2.1]hept-2-ene, 5-Cyano-2-norbornene
Formula:
C₈H₉N MW: 119.17 g/mol Melting Pt: 10 °C Density: 1.01 (20 °C) Flash Pt: 66 °C |
MDL Number:
MFCD00167563 CAS Number: 95-11-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...