5-Norbornen-2-yl acetate (endo- and exo- mixture) ≥98.0%

Supplier: TCI America

Synonyms: 5-Norbornene-2-yl acetate, Bicyclo[2.2.1]hept-5-en-2-yl Acetate, Acetic Acid Bicyclo[2.2.1]hept-5-en-2-yl Ester, Acetic Acid 5-Norbornen-2-yl Ester

N0895-100G N0895-25G
TCN0895-100GEA 363.4 CAD
TCN0895-100G TCN0895-25G
5-Norbornen-2-yl acetate (endo- and exo- mixture) ≥98.0%
5-Norbornen-2-yl acetate (endo- and exo- mixture)

CAS Number: 6143-29-9
MDL Number: MFCD00167565
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 73
Flash Point (°C): 62
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: Acetic Acid 5-Norbornen-2-yl Ester, Bicyclo[2.2.1]hept-5-en-2-yl Acetate, Acetic Acid Bicyclo[2.2.1]hept-5-en-2-yl Ester

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 73 °C
Density: 1.05 (20 °C)
Flash Pt: 62 °C
MDL Number: MFCD00167565
CAS Number: 6143-29-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ee60c1cdb3d669ae647a85294465b38122d7d6e&vid=0ee60c1cdb3d669ae647a85294465b38122d7d6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR