2-Methoxy-4-nitrophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

N0892-25G N0892-5G
TCN0892-25GEA 347.6 CAD
TCN0892-25G TCN0892-5G
2-Methoxy-4-nitrophenol ≥98.0% (by GC, titration analysis)
2-Methoxy-4-nitrophenol

CAS Number: 3251-56-7
MDL Number: MFCD00012143
Molecular Formula: C7H7NO4
Molecular Weight: 169.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 103


Other Product Names / Synonyms: 2-Methoxy-4-nitrophenol

Formula: C₇H₇NO₄
Melting Pt: 103 °C
MDL Number: MFCD00012143
CAS Number: 3251-56-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42bbf64a07179e730ee1d5ca5e2a4c61608d45a8&vid=42bbf64a07179e730ee1d5ca5e2a4c61608d45a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR