1-Nitro-3-(trifluoromethoxy)benzene ≥98.0% (by GC)

Supplier: TCI America

N0884-25G N0884-5G
TCN0884-25GEA 269.39 CAD
TCN0884-25G TCN0884-5G
1-Nitro-3-(trifluoromethoxy)benzene ≥98.0% (by GC)
1-Nitro-3-(trifluoromethoxy)benzene

CAS Number: 2995-45-1
MDL Number: MFCD00041011
Molecular Formula: C7H4F3NO3
Molecular Weight: 207.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Specific Gravity (20/20): 1.45

Formula: C₇H₄F₃NO₃
MW: 207.11 g/mol
Boiling Pt: 96 °C
Density: 1.45 (20 °C)
MDL Number: MFCD00041011
CAS Number: 2995-45-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a2748fdf7ff4771eb14cab83a609c34a6e6ad89&vid=5a2748fdf7ff4771eb14cab83a609c34a6e6ad89&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR