Naphthol AS-BI-β-D-Glucuronide ≥98.0% (by HPLC, total nitrogen) for biochemical research

Supplier: TCI America

N0882-100MG N0882-25MG
TCN0882-100MGEA 499.12 CAD
TCN0882-100MG TCN0882-25MG
Naphthol AS-BI-β-D-Glucuronide ≥98.0% (by HPLC, total nitrogen) for biochemical research
Naphthol AS-BI-beta-D-Glucuronide

[for Biochemical Research]
CAS Number: 37-87-6
MDL Number: MFCD00067162
Molecular Formula: C24H22BrNO9
Molecular Weight: 548.34
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Storage Temperature: <0°C


Product Categories: Bioscience, Glucuronidase Substrates
Other Product Names / Synonyms: 6-Bromo-3[[(2-methoxyphenyl)amino]carbonyl]-2-naphthyl beta-D-Glucopyranosiduronic Acid

Formula: C₂₄H₂₂BrNO₉
MDL Number: MFCD00067162
CAS Number: 37-87-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ccf483b589f040eb2c47e17105e3c047c8f8aff&vid=3497a272047299162ec7f0d08dd0121361c2b01c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR