2-Naphthalenemethanol ≥98.0% (by GC)
Supplier: TCI America
N0881-25G
N0881-5G
TCN0881-25GEA
511.92
CAD
TCN0881-25G
TCN0881-5G
2-Naphthalenemethanol ≥98.0% (by GC)
2-Naphthalenemethanol
CAS Number: 1592-38-7
MDL Number: MFCD00004124
Molecular Formula: C11H10O
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 178
Melting point (°C): 80
Other Product Names / Synonyms: 2-(Hydroxymethyl)naphthalene
Formula:
C₁₁H₁₀O MW: 158.2000 g/mol Boiling Pt: 178 °C Melting Pt: 80 °C |
MDL Number:
MFCD00004124 CAS Number: 1592-38-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...