3-Nitroanthranilic acid ≥97.0%

Supplier: TCI America

Synonyms: 2-Amino-3-nitrobenzoic acid

N0879-25G N0879-5G
TCN0879-25GEA 715.74 CAD
TCN0879-25G TCN0879-5G
3-Nitroanthranilic acid ≥97.0%
3-Nitroanthranilic acid

CAS Number: 606-18-8
MDL Number: MFCD00024261
Molecular Formula: C7H6N2O4
Molecular Weight: 182.14
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 209


Other Product Names / Synonyms: 2-Amino-3-nitrobenzoic Acid

Formula: C₇H₆N₂O₄
Melting Pt: 209 °C
MDL Number: MFCD00024261
CAS Number: 606-18-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e665e8f05166b804deebb9bd9d1ef12b8649523f&vid=e665e8f05166b804deebb9bd9d1ef12b8649523f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR