4-Nitro-L-phenylalanine methyl ester hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
H-Phe(4-NO2)-OMe.HCl, H-Phe(4-NO₂)-OMe·HCl
N0878-1G
N0878-5G
TCN0878-1GEA
36.97
CAD
TCN0878-1G
TCN0878-5G
4-Nitro-L-phenylalanine methyl ester hydrochloride ≥98.0%
4-Nitro-L-phenylalanine methyl ester hydrochloride
CAS Number: 17193-40-7
MDL Number: MFCD00135754
Molecular Formula: C10H12N2O4
Molecular Weight: 260.67
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 224
Specific rotation [a]20/D: 8 deg (C=1, H2O)
Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Phe(4-NO2)-OMe.HCl
Formula:
C₁₀H₁₂N₂O₄ MW: 260.67 g/mol Melting Pt: 224 °C |
MDL Number:
MFCD00135754 CAS Number: 17193-40-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...