4-Nitrobenzyl acetoacetate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4-Nitrobenzyl 3-oxobutanoate, Acetoacetic Acid 4-Nitrobenzyl Ester

N0875-25G N0875-5G
TCN0875-25GEA 107.44 CAD
TCN0875-25G TCN0875-5G
4-Nitrobenzyl acetoacetate ≥98.0% (by HPLC)
4-Nitrobenzyl acetoacetate

CAS Number: 61312-84-3
MDL Number: MFCD00799237
Molecular Formula: C11H11NO5
Molecular Weight: 237.21
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 44
Lambda max.: 267 nm (CH2Cl2)


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: Acetoacetic Acid 4-Nitrobenzyl Ester

Formula: C₁₁H₁₁NO₅
MW: 237.21 g/mol
Melting Pt: 44 °C
MDL Number: MFCD00799237
CAS Number: 61312-84-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5eb7f16e3d45e77909108353c190c0091aa1c4c&vid=b5eb7f16e3d45e77909108353c190c0091aa1c4c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR