Nobiletin ≥95.0% (by HPLC)
Supplier: TCI America
N0871-100MG
N0871-10MG
TCN0871-100MGEA
1369.86
CAD
TCN0871-100MG
TCN0871-10MG
Nobiletin ≥95.0% (by HPLC)
Nobiletin
CAS Number: 478-01-3
MDL Number: MFCD03273560
Molecular Formula: C21H22O8
Molecular Weight: 402.40
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Melting point (°C): 138
Storage Temperature: <0°C
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 3',4',5,6,7,8-Hexamethoxyflavone
Formula:
C₂₁H₂₂O₈ Melting Pt: 138 °C |
MDL Number:
MFCD03273560 CAS Number: 478-01-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...