4-Nitro-1H-pyrazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

N0862-25G N0862-5G
TCN0862-25GEA 67.53 CAD
TCN0862-25G TCN0862-5G
4-Nitro-1H-pyrazole ≥98.0% (by GC, titration analysis)
4-Nitro-1H-pyrazole

CAS Number: 2075-46-9
MDL Number: MFCD00159626
Molecular Formula: C3H3N3O2
Molecular Weight: 113.08
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 323
Melting point (°C): 162
Lambda max.: 317 nm (EtOH)

Formula: C₃H₃N₃O₂
MW: 113.0800 g/mol
Boiling Pt: 323 °C
Melting Pt: 162 °C
MDL Number: MFCD00159626
CAS Number: 2075-46-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db6341a96bcfdc99a388c2e3c94c510730c0a7c2&vid=db6341a96bcfdc99a388c2e3c94c510730c0a7c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR