Neu5Ac[1Me,4789Ac]α(2-3)Gal[246Bz]-β-MP ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: 4-Methoxyphenyl 2,4,6-Tri-O-benzoyl-3-O-(methyl 5-Acetamido-4,7,8,9-tetra-O-acetyl-3,5-dideoxy-D-glycero-α-D-galacto-2-nonulopyranosonate)-β-D-galactopyranoside

N0846-1G N0846-200MG
TCN0846-1GEA 2038.04 CAD
TCN0846-1G TCN0846-200MG
Neu5Ac[1Me,4789Ac]α(2-3)Gal[246Bz]-β-MP ≥95.0% (by HPLC)
Neu5Ac[1Me,4789Ac]alpha(2-3)Gal[246Bz]-beta-MP

Molecular Weight: 1072.04
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Storage Temperature: 0-10°C
Molecular Formula: C54H57NO22


Other Product Names / Synonyms: 4-Methoxyphenyl 2,4,6-Tri-O-benzoyl-3-O-(methyl 5-Acetamido-4,7,8,9-tetra-O-acetyl-3,5-dideoxy-D-glycero-alpha-D-galacto-2-nonulopyranosonate)-beta-D-galactopyranoside

Formula: C₅₄H₅₇NO₂₂
MW: 1072.04 g/mol
CAS Number: 1858223-85-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6b9995e4dfe0adaecb56e2762ccfc80c8a4b768&vid=c6b9995e4dfe0adaecb56e2762ccfc80c8a4b768&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR