4-Nitrobenzyl-(2S,4S)-2-(dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylate ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (2S,4S)-4-Nitrobenzyl 2-(dimethylcarbamoyl)-4-mercaptopyrrolidine-1-carboxylate, (2S,4S)-2-(Dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylic Acid 4-Nitrobenzyl Ester, 4-Nitrobenzyl (2S,4S)-2-(Dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylate

N0828-1G N0828-5G
TCN0828-1GEA 261.17 CAD
TCN0828-1G TCN0828-5G
4-Nitrobenzyl-(2S,4S)-2-(dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylate ≥95.0% (by HPLC, titration analysis)
4-Nitrobenzyl-(2S,4S)-2-(dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylate

CAS Number: 96034-64-9
MDL Number: MFCD08063348
Molecular Formula: C15H19N3O5S
Molecular Weight: 353.39
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Melting point (°C): 119
Specific rotation [a]20/D: 9 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (2S,4S)-2-(Dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylic Acid 4-Nitrobenzyl Ester

Formula: C₁₅H₁₉N₃O₅S
MW: 353.39 g/mol
Melting Pt: 119 °C
MDL Number: MFCD08063348
CAS Number: 96034-64-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3abc1b3177ab43efb22fe97481e4f3583139f3e&vid=f3abc1b3177ab43efb22fe97481e4f3583139f3e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR