Neu5Ac-α(2-6)Gal-β-MP glycoside ≥95.0% (by HPLC)

Supplier: TCI America

N0792-10MG N0792-50MG
TCN0792-10MGEA 500.86 CAD
TCN0792-10MG TCN0792-50MG
Neu5Ac-α(2-6)Gal-β-MP glycoside ≥95.0% (by HPLC)
Neu5Ac-alpha(2-6)Gal-beta-MP glycoside

Molecular Weight: 577.54
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Storage Temperature: <0°C
Molecular Formula: C24H35NO15

Formula: C₂₄H₃₅NO₁₅
CAS Number: 1984814-41-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d157152c9967632254e99d3ecce634f61a5af8b&vid=6d157152c9967632254e99d3ecce634f61a5af8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR