2-Nitro-4-(trifluoromethyl)phenol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
N0779-1G
N0779-5G
TCN0779-1GEA
74.26
CAD
TCN0779-1G
TCN0779-5G
2-Nitro-4-(trifluoromethyl)phenol ≥98.0% (by GC, titration analysis)
2-Nitro-4-(trifluoromethyl)phenol
CAS Number: 400-99-7
MDL Number: MFCD00009791
Molecular Formula: C7H4F3NO3
Molecular Weight: 207.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 94
Flash Point (°C): 95
Specific Gravity (20/20): 1.52
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Hydroxy-3-nitrobenzotrifluoride
Formula:
C₇H₄F₃NO₃ Boiling Pt: 94 °C Density: 1.52 (20 °C) Flash Pt: 95 °C |
MDL Number:
MFCD00009791 CAS Number: 400-99-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...