5-Norbornene-2-carboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
N0667-25G
N0667-5G
TCN0667-25GEA
92.59
CAD
TCN0667-25G
TCN0667-5G
5-Norbornene-2-carboxylic acid ≥98.0% (by GC, titration analysis)
5-Norbornene-2-carboxylic acid
CAS Number: 120-74-1
MDL Number: MFCD00085356
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.13
Other Product Names / Synonyms: Bicyclo[2.2.1]hept-5-ene-2-carboxylic Acid
Formula:
C₈H₁₀O₂ Density: 1.13 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00085356 CAS Number: 120-74-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...