1-Naphthalenemethanol ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Naphthalen-1-ylmethanol

N0633-25G N0633-5G
TCN0633-25GEA 229.91 CAD
TCN0633-25G TCN0633-5G
1-Naphthalenemethanol ≥95.0% (by GC)
1-Naphthalenemethanol

CAS Number: 4780-79-4
MDL Number: MFCD00004044
Molecular Formula: C11H10O
Molecular Weight: 158.20
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 301
Melting point (°C): 62


Other Product Names / Synonyms: 1-(Hydroxymethyl)naphthalene

Formula: C₁₁H₁₀O
MW: 158.20 g/mol
Boiling Pt: 301 °C
Melting Pt: 62 °C
MDL Number: MFCD00004044
CAS Number: 4780-79-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04e4c54141a15fb393a0088a227c8b68b75e3f8c&vid=04e4c54141a15fb393a0088a227c8b68b75e3f8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR