Nitrilotris(methylenephosphonic acid) ∼50% ∼2.2 mol/L in water

Supplier: TCI America

Synonyms: Tris(phosphonomethyl)amine, Nitrilotri(methylphosphonic acid)

N0474-25ML N0474-500ML
TCN0474-25MLEA 77.14 CAD
TCN0474-25ML TCN0474-500ML
Nitrilotris(methylenephosphonic acid) ∼50% ∼2.2 mol/L in water
Nitrilotris(methylenephosphonic acid)

CAS Number: 6419-19-8
MDL Number: MFCD00002138
Molecular Formula: C3H12NO9P3
Molecular Weight: 299.05
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.33


Other Product Names / Synonyms: Tris(phosphonomethyl)amine

Formula: C₃H₁₂NO₉P₃
MW: 299.05 g/mol
Density: 1.33 (20 °C)
MDL Number: MFCD00002138
CAS Number: 6419-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f879a7a408bf2478d3d1bcd5ebe9035169ce59f&vid=99093dac6b82df2d03b9bec3826f94ed77370ead&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR