6-Nitro-m-toluic acid ≥98.0%

Supplier: TCI America

Synonyms: 2-Nitro-5-methylbenzoic acid, 5-Methyl-2-nitrobenzoic acid

N0470-25G N0470-5G
TCN0470-25GEA 67.78 CAD
TCN0470-25G TCN0470-5G
6-Nitro-m-toluic acid ≥98.0%
6-Nitro-m-toluic acid

CAS Number: 3113-72-2
MDL Number: MFCD00007368
Molecular Formula: C8H7NO4
Molecular Weight: 181.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 136


Other Product Names / Synonyms: 6-Nitro-m-toluic Acid

Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 364
Melting Pt: 136 °C
MDL Number: MFCD00007368
CAS Number: 3113-72-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89064f7a63c40d678652cc7d7a231a16cdf56e2d&vid=e504b7318ff49846fb36421e37fa6d1246068544&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR