4-Nitroquinoline-N-oxide ≥98.0%

Supplier: TCI America

Synonyms: 4-Nitroquinoline 1-oxide, 4-Nitroquinoline N-oxide

N0250-1G N0250-5G
TCN0250-1GEA 168.78 CAD
TCN0250-1G TCN0250-5G
4-Nitroquinoline-N-oxide ≥98.0%
4-Nitroquinoline N-oxide

CAS Number: 56-57-5
MDL Number: MFCD00006738
Molecular Formula: C9H6N2O3
Molecular Weight: 190.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 156

Formula: C₉H₆N₂O₃
Melting Pt: 156 °C
MDL Number: MFCD00006738
CAS Number: 56-57-5
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09ce7fb423ee109eb32349fe82806b04e748ef16&vid=09ce7fb423ee109eb32349fe82806b04e748ef16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR