2-(3-Methoxyphenyl)ethanol ≥98.0%

Supplier: TCI America

M2658-1G M2658-5G
TCM2658-1GEA 88.48 CAD
TCM2658-1G TCM2658-5G
2-(3-Methoxyphenyl)ethanol ≥98.0%
2-(3-Methoxyphenyl)ethanol

CAS Number: 5020-41-7
MDL Number: MFCD00002893
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 113
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: 3-(2-Hydroxyethyl)anisole, 2-(3-Methoxyphenyl)ethanol

Formula: C₉H₁₂O₂
Boiling Pt: 140 °C
Density: 1.09 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00002893
CAS Number: 5020-41-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5bec231d7c4245e3be2c02d12b660104b8e04843&vid=5bec231d7c4245e3be2c02d12b660104b8e04843&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR