2-(2-Methoxyphenyl)ethanol ≥98.0%
Supplier: TCI America
M2657-1G
M2657-5G
TCM2657-1GEA
67.94
CAD
TCM2657-1G
TCM2657-5G
2-(2-Methoxyphenyl)ethanol ≥98.0%
2-(2-Methoxyphenyl)ethanol
CAS Number: 7417-18-7
MDL Number: MFCD00002889
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 134
Flash Point (°C): 113
Specific Gravity (20/20): 1.09
Other Product Names / Synonyms: 2-(2-Hydroxyethyl)anisole, 2-(2-Methoxyphenyl)ethanol
Formula:
C₉H₁₂O₂ Boiling Pt: 134 °C Density: 1.09 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00004512 CAS Number: 7417-18-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...