2-Methoxy-5-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M2650-1G M2650-5G
TCM2650-1GEA 53.72 CAD
TCM2650-1G TCM2650-5G
2-Methoxy-5-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Methoxy-5-pyridineboronic acid pinacol ester

CAS Number: 445264-61-9
MDL Number: MFCD05663858
Molecular Formula: C12H18BNO3
Molecular Weight: 235.09
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 102
Melting point (°C): 53


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 2-(6-Methoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-Methoxy-3-pyridylboronic Acid Pinacol Ester

Formula: C₁₂H₁₈BNO₃
Boiling Pt: 102 °C
Melting Pt: 53 °C
MDL Number: MFCD05663858
CAS Number: 445264-61-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=326e515c8a05321aa32bac456941c1631030308e&vid=326e515c8a05321aa32bac456941c1631030308e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR