Methyl-6-chloropicolinate ≥98.0%
Supplier: TCI America
M2640-1G
M2640-5G
TCM2640-1GEA
26.86
CAD
TCM2640-1G
TCM2640-5G
Methyl-6-chloropicolinate ≥98.0%
Methyl 6-chloropicolinate
CAS Number: 6636-55-1
MDL Number: MFCD00155391
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 95
Other Product Names / Synonyms: 6-Chloro-2-pyridinecarboxylic Acid Methyl Ester, Methyl 6-Chloropicolinate, 6-Chloropicolinic Acid Methyl Ester
Formula:
C₇H₆ClNO₂ Melting Pt: 95 °C |
MDL Number:
MFCD00155391 CAS Number: 6636-55-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...