(+)-Menthyl acetate ≥98.0% (by GC)

Supplier: TCI America

M2631-1ML M2631-5ML
TCM2631-1MLEA 84.37 CAD
TCM2631-1ML TCM2631-5ML
(+)-Menthyl acetate ≥98.0% (by GC)
(+)-Menthyl acetate

CAS Number: 5157-89-1
MDL Number: MFCD00062977
Molecular Formula: C12H22O2
Molecular Weight: 198.31
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 229
Flash Point (°C): 77
Specific Gravity (20/20): 0.93
Specific rotation [a]20/D: 77 deg (neat)


Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: (+)-Acetoxy-p-menthane

Formula: C₁₂H₂₂O₂
Boiling Pt: 229 °C
Density: 0.93 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00062977
CAS Number: 5157-89-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79d3139aec8344499c9bc636b4309fc3033e42c8&vid=79d3139aec8344499c9bc636b4309fc3033e42c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR