Methyl thiophene-3-carboxylate ≥97.0% (by GC)
Supplier: TCI America
M2627-25G
M2627-5G
TCM2627-25GEA
309.68
CAD
TCM2627-25G
TCM2627-5G
Methyl thiophene-3-carboxylate ≥97.0% (by GC)
Methyl thiophene-3-carboxylate
CAS Number: 22913-26-4
MDL Number: MFCD06203810
Molecular Formula: C6H6O2S
Molecular Weight: 142.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 91
Flash Point (°C): 77
Specific Gravity (20/20): 1.23
Lambda max.: 239 nm (Hexane)
Other Product Names / Synonyms: Thiophene-3-carboxylic Acid Methyl Ester
Formula:
C₆H₆O₂S Boiling Pt: 91 °C Density: 1.23 (20 °C) Flash Pt: 77 °C |
MDL Number:
MFCD06203810 CAS Number: 22913-26-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...