2-Methylquinoline-6-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

M2625-25G M2625-5G
TCM2625-25GEA 150.42 CAD
TCM2625-25G TCM2625-5G
2-Methylquinoline-6-carboxylic acid ≥98.0% (by HPLC, titration analysis)
2-Methylquinoline-6-carboxylic acid

CAS Number: 635-80-3
MDL Number: MFCD00748504
Molecular Formula: C11H9NO2
Molecular Weight: 187.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 263

Formula: C₁₁H₉NO₂
MW: 187.19 g/mol
Melting Pt: 263 °C
MDL Number: MFCD00748504
CAS Number: 635-80-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60728a0fe4a486986c6f37d13bd5b173beacaff2&vid=60728a0fe4a486986c6f37d13bd5b173beacaff2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR