2-Hydroxyethylmesitylene ≥98.0%

Supplier: TCI America

Synonyms: 1-(2-Hydroxyethyl)-2,4,6-trimethylbenzene, 2-(2,4,6-Trimethylphenyl)ethanol, 2-Hydroxyethylmesitylene, 2,4,6-Trimethylphenethyl alcohol, 2-Mesitylethanol

M2624-1G M2624-5G
TCM2624-1GEA 41.08 CAD
TCM2624-1G TCM2624-5G
2-Hydroxyethylmesitylene ≥98.0%
2-Hydroxyethylmesitylene

CAS Number: 6950-92-1
MDL Number: MFCD00020609
Molecular Formula: C11H16O
Molecular Weight: 164.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 152
Melting point (°C): 80


Other Product Names / Synonyms: 2-Hydroxyethylmesitylene, 2,4,6-Trimethylphenethyl Alcohol

Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Boiling Pt: 152 °C
Melting Pt: 80 °C
MDL Number: MFCD00020609
CAS Number: 6950-92-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c4ba7c27025996822ed5d2d90161309f7f8f00b&vid=4c4ba7c27025996822ed5d2d90161309f7f8f00b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR