2-Methyl-4-quinolinol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M2620-25G M2620-5G
TCM2620-25GEA 793.16 CAD
TCM2620-25G TCM2620-5G
2-Methyl-4-quinolinol ≥98.0% (by GC, titration analysis)
2-Methyl-4-quinolinol

CAS Number: 607-67-0
MDL Number: MFCD00006758
Molecular Formula: C10H9NO
Molecular Weight: 159.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 235
Lambda max.: 317 nm (CHCl3)


Other Product Names / Synonyms: 4-Hydroxy-2-methylquinoline

Formula: C₁₀H₉NO
Melting Pt: 235 °C
MDL Number: MFCD00006758
CAS Number: 607-67-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c874d4885e646549ec295bf205dbfe4cbf0f3ad8&vid=c874d4885e646549ec295bf205dbfe4cbf0f3ad8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR