Methyl-2,6-dichloronicotinate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 2,6-dichloronicotinate

M2616-1G M2616-5G
TCM2616-1GEA 69.52 CAD
TCM2616-1G TCM2616-5G
Methyl-2,6-dichloronicotinate ≥98.0% (by GC)
Methyl 2,6-dichloronicotinate

CAS Number: 65515-28-8
MDL Number: MFCD07369794
Molecular Formula: C7H5Cl2NO2
Molecular Weight: 206.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 59
Lambda max.: 276 nm


Other Product Names / Synonyms: 2,6-Dichloronicotinic Acid Methyl Ester, Methyl 2,6-Dichloropyridine-3-carboxylate, 2,6-Dichloropyridine-3-carboxylic Acid Methyl Ester

Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
Melting Pt: 59 °C
MDL Number: MFCD07369794
CAS Number: 65515-28-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc3073f293ef6168feba0f1e17840cb71c8b6a12&vid=bc3073f293ef6168feba0f1e17840cb71c8b6a12&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR