4-Methoxybenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
M2612-1G
M2612-5G
TCM2612-1GEA
170.64
CAD
TCM2612-1G
TCM2612-5G
4-Methoxybenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Methoxybenzamidine hydrochloride
CAS Number: 51721-68-7
MDL Number: MFCD00466114
Molecular Formula: C8H10N2O
Molecular Weight: 186.64
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 221
Lambda max.: 261 nm (H2O)
Formula:
C₈H₁₀N₂O Melting Pt: 221 °C |
MDL Number:
MFCD00466114 CAS Number: 51721-68-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...