Methyl-2-aminothiazole-5-carboxylate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 2-aminothiazole-5-carboxylate, 2-Aminothiazole-5-carboxylic acid methyll ester

M2598-1G M2598-5G
TCM2598-1GEA 90.06 CAD
TCM2598-1G TCM2598-5G
Methyl-2-aminothiazole-5-carboxylate ≥98.0% (by GC)
Methyl-2-aminothiazole-5-carboxylate

CAS Number: 6633-61-0
MDL Number: MFCD03788562
Molecular Formula: C5H6N2O2S
Molecular Weight: 158.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 191
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Aminothiazole-5-carboxylic Acid Methyl Ester

Formula: C₅H₆N₂O₂S
MW: 158.1800 g/mol
Melting Pt: 191 °C
MDL Number: MFCD03788562
CAS Number: 6633-61-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f695bdc9aca96d3ea7a76de287f128f5ab969c3&vid=8f695bdc9aca96d3ea7a76de287f128f5ab969c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR