Methyl-3-bromothiophene-2-carboxylate ≥98.0%
Supplier: TCI America
M2590-1G
M2590-5G
TCM2590-1GEA
110.6
CAD
TCM2590-1G
TCM2590-5G
Methyl-3-bromothiophene-2-carboxylate ≥98.0%
Methyl-3-bromothiophene-2-carboxylate
CAS Number: 26137-08-6
MDL Number: MFCD00173839
Molecular Formula: C6H5BrO2S
Molecular Weight: 221.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 127
Melting point (°C): 49
Other Product Names / Synonyms: 3-Bromothiophene-2-carboxylic Acid Methyl Ester
Formula:
C₆H₅BrO₂S Boiling Pt: 127 °C Melting Pt: 49 °C |
MDL Number:
MFCD00173839 CAS Number: 26137-08-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...