Methyl-4-bromo-1H-pyrrole-2-carboxylate ≥98.0% (by GC)

Supplier: TCI America

M2583-1G M2583-5G
TCM2583-1GEA 55.3 CAD
TCM2583-1G TCM2583-5G
Methyl-4-bromo-1H-pyrrole-2-carboxylate ≥98.0% (by GC)
Methyl-4-bromo-1H-pyrrole-2-carboxylate

CAS Number: 934-05-4
MDL Number: MFCD00832855
Molecular Formula: C6H6BrNO2
Molecular Weight: 204.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 103
Lambda max.: 271 nm (MeOH)


Other Product Names / Synonyms: 4-Bromopyrrole-2-carboxylic Acid Methyl Ester

Formula: C₆H₆BrNO₂
Melting Pt: 103 °C
MDL Number: MFCD00832855
CAS Number: 934-05-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2dd034ea0ff905311455375e2e6a4070ff0653ab&vid=2dd034ea0ff905311455375e2e6a4070ff0653ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR