6-(Methoxycarbonyl)picolinic acid ≥97.0%
Supplier: TCI America
Synonyms:
2,6-Pyridinedicarboxylic acid monomethyl ester, Dipicolinic acid monomethyl ester, Monomethyl 2,6-Pyridinedicarboxylate, Monomethyl dipicolinate
CAS Number: 7170-36-7
MDL Number: MFCD00806109
Molecular Formula: C8H7NO4
Molecular Weight: 181.15
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 153
Other Product Names / Synonyms: 2,6-Pyridinedicarboxylic Acid Monomethyl Ester, Monomethyl Dipicolinate, Dipicolinic Acid Monomethyl Ester
Formula:
C8H7NO4 MW: 181.1455 g/mol Melting Pt: 153 °C |
MDL Number:
MFCD00806109 CAS Number: 7170-36-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...