4-Methoxyphenylurea ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

M2575-1G M2575-5G
TCM2575-1GEA 79 CAD
TCM2575-1G TCM2575-5G
4-Methoxyphenylurea ≥98.0% (by HPLC, total nitrogen)
4-Methoxyphenylurea

CAS Number: 1566-42-3
MDL Number: MFCD00014789
Molecular Formula: C8H10N2O2
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 162
Lambda max.: 289 nm (EtOH)

Formula: C₈H₁₀N₂O₂
Melting Pt: 162 °C
MDL Number: MFCD00014789
CAS Number: 1566-42-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=599a666f64c87494b4472abb7e1d82cde2d3ec02&vid=599a666f64c87494b4472abb7e1d82cde2d3ec02&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR