6-Methyl-2-(p-tolylamino)-4H-3,1-benzoxazin-4-one ≥95.0% (by HPLC)

Supplier: TCI America

M2572-10MG M2572-50MG
TCM2572-10MGEA 110.76 CAD
TCM2572-10MG TCM2572-50MG
6-Methyl-2-(p-tolylamino)-4H-3,1-benzoxazin-4-one ≥95.0% (by HPLC)
6-Methyl-2-(p-tolylamino)-4H-3,1-benzoxazin-4-one

CAS Number: 86672-58-4
MDL Number: MFCD03272508
Molecular Formula: C16H14N2O2
Molecular Weight: 266.30
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Melting point (°C): 215
Storage Temperature: >-20°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research)
Other Product Names / Synonyms: URB-754, 6-Methyl-2-(p-tolylamino)-4H-benzo[d][1,3]oxazin-4-one

Formula: C₁₆H₁₄N₂O₂
Melting Pt: 215 °C
MDL Number: MFCD03272508
CAS Number: 86672-58-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f51c0e67bdcebcfd59fa80602d7d2ab2d7309e62&vid=f51c0e67bdcebcfd59fa80602d7d2ab2d7309e62&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR