5-Nitro-o-toluonitrile ≥98.0%

Supplier: TCI America

Synonyms: 2-Methyl-5-nitrobenzonitrile

M2570-25G M2570-5G
TCM2570-25GEA 167.01 CAD
TCM2570-25G TCM2570-5G
5-Nitro-o-toluonitrile ≥98.0%
5-Nitro-o-toluonitrile

CAS Number: 939-83-3
MDL Number: MFCD00017040
Molecular Formula: C8H6N2O2
Molecular Weight: 162.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 175
Melting point (°C): 106


Other Product Names / Synonyms: 5-Nitro-o-tolunitrile

Formula: C₈H₆N₂O₂
Boiling Pt: 175 °C
Melting Pt: 106 °C
MDL Number: MFCD00017040
CAS Number: 939-83-3
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6d6cdd076ed6e8e822fbc812d6ae472a8ff3016&vid=e6d6cdd076ed6e8e822fbc812d6ae472a8ff3016&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR