2-Methylnaphth[2,1-d]oxazole ≥98.0%

Supplier: TCI America

Synonyms: 2-Methylnaphtho[2,1-d]oxazole

M2559-1G M2559-5G
TCM2559-1GEA 84.37 CAD
TCM2559-1G TCM2559-5G
2-Methylnaphth[2,1-d]oxazole ≥98.0%
2-Methylnaphth[2,1-d]oxazole

CAS Number: 20686-65-1
MDL Number: MFCD00022185
Molecular Formula: C12H9NO
Molecular Weight: 183.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 302
Melting point (°C): 41
Lambda max.: 283 nm (Octane)

Formula: C₁₂H₉NO
MW: 183.21 g/mol
Boiling Pt: 302 °C
Melting Pt: 41 °C
MDL Number: MFCD00022185
CAS Number: 20686-65-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1967c19a5d55f44320cc3d8dc127da36c3f9efe&vid=d1967c19a5d55f44320cc3d8dc127da36c3f9efe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR