1-Methanesulfonyl-2-imidazolidinone ≥98.0% (by GC)

Supplier: TCI America

M2553-25G M2553-5G
TCM2553-25GEA 116.92 CAD
TCM2553-25G TCM2553-5G
1-Methanesulfonyl-2-imidazolidinone ≥98.0% (by GC)
1-Methanesulfonyl-2-imidazolidinone

CAS Number: 41730-79-4
MDL Number: MFCD07787278
Molecular Formula: C4H8N2O3S
Molecular Weight: 164.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 193

Formula: C₄H₈N₂O₃S
Melting Pt: 193 °C
MDL Number: MFCD07787278
CAS Number: 41730-79-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bef2d8a1d2e219de22162e5fab797e17f3df537&vid=4bef2d8a1d2e219de22162e5fab797e17f3df537&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR