N-Methyl-2-(4-methoxyphenyl)ethylamine ≥95.0%

Supplier: TCI America

Synonyms: 4-[2-(Methylamino)ethyl]anisole, 4-Methoxy-N-methylphenethylamine

M2551-200MG M2551-20MG
TCM2551-200MGEA 322.32 CAD
TCM2551-200MG TCM2551-20MG
N-Methyl-2-(4-methoxyphenyl)ethylamine ≥95.0%
N-Methyl-2-(4-methoxyphenyl)ethylamine

CAS Number: 4091-50-3
MDL Number: MFCD00870496
Molecular Formula: C10H15NO
Molecular Weight: 165.24
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 83
Specific Gravity (20/20): 1.01


Product Categories: Bioscience, Reagents for Pain & Itch Models, Reagents for Inflammation & Immunity Disorder Models
Other Product Names / Synonyms: 4-Methoxy-N-methylphenethylamine, 4-[2-(Methylamino)ethyl]anisole

Formula: C₁₀H₁₅NO
Boiling Pt: 83 °C
Density: 1.01 (20 °C)
MDL Number: MFCD00870496
CAS Number: 4091-50-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edcd0009d340701ff86955d74917067cddc5d6bf&vid=edcd0009d340701ff86955d74917067cddc5d6bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR