Methyl-2-methyl-4-nitrobenzoate ≥98.0% (by GC)
Supplier: TCI America
M2545-25G
M2545-5G
TCM2545-25GEA
410.8
CAD
TCM2545-25G
TCM2545-5G
Methyl-2-methyl-4-nitrobenzoate ≥98.0% (by GC)
Methyl 2-methyl-4-nitrobenzoate
CAS Number: 62621-09-4
MDL Number: MFCD03618407
Molecular Formula: C9H9NO4
Molecular Weight: 195.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 76
Other Product Names / Synonyms: 2-Methyl-4-nitrobenzoic Acid Methyl Ester, Methyl 4-Nitro-o-toluate, 4-Nitro-o-toluic Acid Methyl Ester
Formula:
C₉H₉NO₄ Melting Pt: 76 °C |
MDL Number:
MFCD03618407 CAS Number: 62621-09-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...