Methyl-5-methyl-2-nitrobenzoate ≥98.0%
Supplier: TCI America
Synonyms:
6-Nitro-m-toluic acid methyll ester, Methyl 6-Nitro-m-toluate, Methyl 5-methyl-2-nitrobenzoate, 5-Methyl-2-nitrobenzoic acid methyll ester
M2544-25G
M2544-5G
TCM2544-25GEA
189.6
CAD
TCM2544-25G
TCM2544-5G
Methyl-5-methyl-2-nitrobenzoate ≥98.0%
Methyl 5-methyl-2-nitrobenzoate
CAS Number: 20587-30-8
MDL Number: MFCD00157275
Molecular Formula: C9H9NO4
Molecular Weight: 195.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 80
Other Product Names / Synonyms: 5-Methyl-2-nitrobenzoic Acid Methyl Ester, Methyl 6-Nitro-m-toluate, 6-Nitro-m-toluic Acid Methyl Ester
Formula:
C₉H₉NO₄ MW: 195.1721 g/mol Melting Pt: 80 °C |
MDL Number:
MFCD00157275 CAS Number: 20587-30-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...