Methyl-2-amino-5-methylbenzoate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M2541-25G M2541-5G
TCM2541-25GEA 370.19 CAD
TCM2541-25G TCM2541-5G
Methyl-2-amino-5-methylbenzoate ≥98.0% (by GC, titration analysis)
Methyl 2-amino-5-methylbenzoate

CAS Number: 18595-16-9
MDL Number: MFCD00661547
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 63


Other Product Names / Synonyms: 2-Amino-5-methylbenzoic Acid Methyl Ester, Methyl 6-Amino-m-toluate, 6-Amino-m-toluic Acid Methyl Ester, Methyl 5-Methylanthranilate, 5-Methylanthranilic Acid Methyl Ester

Formula: C₉H₁₁NO₂
Melting Pt: 63 °C
MDL Number: MFCD08234606
CAS Number: 18595-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fff1546a4122cf53fcfbe2363d4a88b067da4c9&vid=0fff1546a4122cf53fcfbe2363d4a88b067da4c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR